Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTATGGGCAGGGCTGGGCCTCA[C/T]GTCTGGGCCAGCGAGAGCTCCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC154 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC154 - coiled-coil domain containing 154 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCN7 - chloride voltage-gated channel 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114331.2 | 2453 | Silent Mutation | ACA,ACG | T,T 781 | NP_001107803.1 | |
NM_001287.5 | 2453 | Silent Mutation | ACA,ACG | T,T 805 | NP_001278.1 | |
XM_011522354.1 | 2453 | Silent Mutation | ACA,ACG | T,T 747 | XP_011520656.1 |