Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTCTCTCTCTTTGCAGATGAC[C/G]GCTTCCCGAAGCCCCCTGAGATTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 140210 | ||||||||||||||||||||
Literature Links: |
HPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPR - haptoglobin-related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020995.3 | 127 | Missense Mutation | CGC,GGC | R,G 33 | NP_066275.3 |
TXNL4B - thioredoxin like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142317.1 | 127 | Intron | NP_001135789.1 | |||
NM_001142318.1 | 127 | Intron | NP_001135790.1 | |||
NM_001324354.1 | 127 | Intron | NP_001311283.1 | |||
NM_001324355.1 | 127 | Intron | NP_001311284.1 | |||
NM_017853.2 | 127 | Intron | NP_060323.1 | |||
XM_017023377.1 | 127 | Intron | XP_016878866.1 |