Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCTCCTGGCCGTCTGCGCCCTC[A/G]TGTCCTTGGCAACCGCTGCACTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613799 MIM: 606800 | ||||||||||||||||||||
Literature Links: |
CCDC40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC40 - coiled-coil domain containing 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAA - glucosidase alpha, acid | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000152.4 | 293 | Missense Mutation | ATG,GTG | M,V 19 | NP_000143.2 | |
NM_001079803.2 | 293 | Missense Mutation | ATG,GTG | M,V 19 | NP_001073271.1 | |
NM_001079804.2 | 293 | Missense Mutation | ATG,GTG | M,V 19 | NP_001073272.1 | |
XM_005257193.2 | 293 | Missense Mutation | ATG,GTG | M,V 19 | XP_005257250.1 | |
XM_005257194.4 | 293 | Missense Mutation | ATG,GTG | M,V 19 | XP_005257251.1 |
MIR1268B - microRNA 1268b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |