Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGGTGGTCATGTCACAGAAGAC[A/G]GGCACAGGCACACTGGGGCCCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614144 MIM: 602521 MIM: 600596 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B9D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B9D1 - B9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK7 - mitogen-activated protein kinase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFAP4 - microfibrillar associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198695.1 | 265 | Silent Mutation | CCC,CCT | P,P 91 | NP_001185624.1 | |
NM_002404.2 | 265 | Silent Mutation | CCC,CCT | P,P 67 | NP_002395.1 |