Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGTCCTCACTTCTCCCCGGAAG[C/T]GGCAGGACTCTTGAAGACCTCAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602076 MIM: 607066 | ||||||||||||||||||||
Literature Links: |
TRPV1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRPV1 - transient receptor potential cation channel subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018727.5 | 2704 | Missense Mutation | ACT,GCT | T,A 835 | NP_061197.4 | |
NM_080704.3 | 2704 | Missense Mutation | ACT,GCT | T,A 835 | NP_542435.2 | |
NM_080705.3 | 2704 | Missense Mutation | ACT,GCT | T,A 835 | NP_542436.2 | |
NM_080706.3 | 2704 | Missense Mutation | ACT,GCT | T,A 835 | NP_542437.2 |
TRPV3 - transient receptor potential cation channel subfamily V member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |