Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGAAGGGAAGGAGGGGTCGGAG[A/G]ACGAGGACGAGGACAACCAGAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608417 | ||||||||||||||||||||
Literature Links: |
C17orf50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf50 - chromosome 17 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145272.3 | 145 | Missense Mutation | AAC,GAC | N,D 34 | NP_660315.2 |
MMP28 - matrix metallopeptidase 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032278.2 | 145 | Intron | NP_001027449.1 | |||
NM_024302.4 | 145 | Intron | NP_077278.1 | |||
NM_032950.3 | 145 | Intron | NP_116568.1 | |||
XM_011525225.1 | 145 | Intron | XP_011523527.1 | |||
XM_011525226.2 | 145 | Intron | XP_011523528.1 | |||
XM_011525227.1 | 145 | Intron | XP_011523529.1 | |||
XM_011525228.1 | 145 | Intron | XP_011523530.1 | |||
XM_011525229.2 | 145 | Intron | XP_011523531.1 | |||
XM_011525230.1 | 145 | Intron | XP_011523532.1 | |||
XM_011525231.1 | 145 | Intron | XP_011523533.1 | |||
XM_011525232.2 | 145 | Intron | XP_011523534.2 | |||
XM_017025061.1 | 145 | Intron | XP_016880550.1 | |||
XM_017025062.1 | 145 | Intron | XP_016880551.1 | |||
XM_017025063.1 | 145 | Intron | XP_016880552.1 | |||
XM_017025064.1 | 145 | Intron | XP_016880553.1 |