Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGGCCCCAGGCCGGCCGTGCC[A/G]TTGACTTGCTGGTGCGAGAGGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603039 MIM: 611418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MNT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MNT - MAX network transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020310.2 | 876 | Silent Mutation | AAC,AAT | N,N 530 | NP_064706.1 | |
XM_011523868.2 | 876 | Silent Mutation | AAC,AAT | N,N 538 | XP_011522170.1 | |
XM_011523869.1 | 876 | Silent Mutation | AAC,AAT | N,N 249 | XP_011522171.1 | |
XM_017024654.1 | 876 | Silent Mutation | AAC,AAT | N,N 249 | XP_016880143.1 |
SGSM2 - small G protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |