Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGTTCCTTGGCCTCCTCCAGCTC[A/G]TGCTGGAGCTTGCGGAACTTGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYH4 - myosin heavy chain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017533.2 | 5970 | Silent Mutation | CAC,CAT | H,H 1905 | NP_060003.2 | |
XM_017024676.1 | 5970 | Silent Mutation | CAC,CAT | H,H 1905 | XP_016880165.1 |
MYHAS - myosin heavy chain gene cluster antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |