Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCCGCAGCGGCCGCTGCCCC[A/C]TGCACTGCAGGTCTGGGGACTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601237 | ||||||||||||||||||||
Literature Links: |
RNF112 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF112 - ring finger protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007148.4 | 343 | Missense Mutation | CAT,CCT | H,P 124 | NP_009079.2 | |
XM_006721571.3 | 343 | Missense Mutation | CAT,CCT | H,P 124 | XP_006721634.1 | |
XM_006721572.3 | 343 | Missense Mutation | CAT,CCT | H,P 124 | XP_006721635.1 | |
XM_006721573.3 | 343 | Missense Mutation | CAT,CCT | H,P 100 | XP_006721636.1 |