Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTCTCACTTTCTTCTTCTCCTT[A/G]AAGCGTCCTGAGTTGGAGACCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603287 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPO - pyridoxamine 5'-phosphate oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR15L - proline rich 15 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024320.3 | 508 | Silent Mutation | TTC,TTT | F,F 76 | NP_077296.1 | |
XM_005257663.4 | 508 | Silent Mutation | TTC,TTT | F,F 76 | XP_005257720.1 | |
XM_005257664.2 | 508 | Silent Mutation | TTC,TTT | F,F 76 | XP_005257721.1 | |
XM_005257665.1 | 508 | Silent Mutation | TTC,TTT | F,F 76 | XP_005257722.1 | |
XM_005257666.4 | 508 | Silent Mutation | TTC,TTT | F,F 76 | XP_005257723.1 |