Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCGCTCCACTTTGCCCAGAATC[C/T]GGCCTTCATACATCGGGAGCACCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AXIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AXIN2 - axin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004655.3 | 3001 | Missense Mutation | CAG,CGG | Q,R 834 | NP_004646.3 | |
XM_011525319.2 | 3001 | Missense Mutation | CAG,CGG | Q,R 834 | XP_011523621.1 | |
XM_011525320.1 | 3001 | Missense Mutation | CAG,CGG | Q,R 834 | XP_011523622.1 | |
XM_011525321.2 | 3001 | Missense Mutation | CAG,CGG | Q,R 834 | XP_011523623.1 | |
XM_017025192.1 | 3001 | Missense Mutation | CAG,CGG | Q,R 834 | XP_016880681.1 | |
XM_017025193.1 | 3001 | Missense Mutation | CAG,CGG | Q,R 769 | XP_016880682.1 | |
XM_017025194.1 | 3001 | Missense Mutation | CAG,CGG | Q,R 769 | XP_016880683.1 |