Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTCCAGATGAACCGACGTCAC[C/T]GGATGCCTGGATATGAGACCATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602539 | ||||||||||||||||||||
Literature Links: |
MAP3K3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K3 - mitogen-activated protein kinase kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002401.3 | 621 | Missense Mutation | CGG,TGG | R,W 24 | NP_002392.2 | |
NM_203351.1 | 621 | Missense Mutation | CGG,TGG | R,W 24 | NP_976226.1 | |
XM_005257376.3 | 621 | Missense Mutation | CGG,TGG | R,W 24 | XP_005257433.1 | |
XM_005257377.3 | 621 | Missense Mutation | CGG,TGG | R,W 24 | XP_005257434.1 | |
XM_005257378.2 | 621 | Missense Mutation | CGG,TGG | R,W 24 | XP_005257435.1 |