Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGATGGTGCCTGCTGCCTGGCT[C/T]GGTGGCTCCTGCAATGGCAAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603060 MIM: 607883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF1C - kinesin family member 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC52A1 - solute carrier family 52 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104577.1 | 1484 | Silent Mutation | CCA,CCG | P,P 257 | NP_001098047.1 | |
NM_017986.3 | 1484 | Silent Mutation | CCA,CCG | P,P 257 | NP_060456.3 | |
XM_011523951.1 | 1484 | Silent Mutation | CCA,CCG | P,P 257 | XP_011522253.1 |