Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCCTTGAAAGCAGAAGAAGTG[C/T]CAGCCCTCAGCTTCCGTCAGATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611463 MIM: 182205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAT2 - spermidine/spermine N1-acetyltransferase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320845.1 | 720 | Nonsense Mutation | TGA,TGG | *,W 233 | NP_001307774.1 | |
NM_001320846.1 | 720 | Nonsense Mutation | TGA,TGG | *,W 199 | NP_001307775.1 | |
NM_001320847.1 | 720 | Nonsense Mutation | TGA,TGG | *,W 120 | NP_001307776.1 | |
NM_133491.4 | 720 | Nonsense Mutation | TGA,TGG | *,W 154 | NP_597998.1 | |
XM_017024073.1 | 720 | UTR 3 | XP_016879562.1 | |||
XM_017024074.1 | 720 | UTR 3 | XP_016879563.1 |
SHBG - sex hormone binding globulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040.4 | 720 | Intron | NP_001031.2 | |||
NM_001146279.2 | 720 | Intron | NP_001139751.1 | |||
NM_001146280.2 | 720 | Intron | NP_001139752.1 | |||
NM_001146281.2 | 720 | Intron | NP_001139753.1 | |||
NM_001289113.1 | 720 | Intron | NP_001276042.1 | |||
NM_001289114.1 | 720 | Intron | NP_001276043.1 | |||
NM_001289115.1 | 720 | Intron | NP_001276044.1 | |||
NM_001289116.1 | 720 | Intron | NP_001276045.1 |