Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCGCCAGGGACCCCCTGGCAGC[C/G]GGAGCCGCGGGTCGAGGTTATGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPHK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPHK1 - sphingosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142601.1 | 373 | UTR 5 | NP_001136073.1 | |||
NM_001142602.1 | 373 | UTR 5 | NP_001136074.1 | |||
NM_021972.3 | 373 | UTR 5 | NP_068807.2 | |||
NM_182965.2 | 373 | Missense Mutation | CCG,CGG | P,R 80 | NP_892010.2 | |
XM_005257766.2 | 373 | UTR 5 | XP_005257823.1 |
UBE2O - ubiquitin conjugating enzyme E2 O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |