Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGAGGTTCCACGGCTGGCTGG[C/T]GTTCCTTCCTGGAATGGAGGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606636 | ||||||||||||||||||||
Literature Links: |
LOC728392 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC728392 - uncharacterized LOC728392 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162371.2 | 4633 | Intron | NP_001155843.1 |
NLRP1 - NLR family pyrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033053.2 | 4633 | Missense Mutation | ACC,GCC | T,A 1360 | NP_001028225.1 | |
NM_014922.4 | 4633 | Intron | NP_055737.1 | |||
NM_033004.3 | 4633 | Intron | NP_127497.1 | |||
NM_033006.3 | 4633 | Intron | NP_127499.1 | |||
NM_033007.3 | 4633 | Intron | NP_127500.1 |