Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAGTGGAAATTCCTGCACAGG[A/T]GGGTATACATAGCTGCCCCCGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100710 MIM: 612308 | ||||||||||||||||||||
Literature Links: |
CHRNB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNB1 - cholinergic receptor nicotinic beta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB4 - zinc finger and BTB domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128833.1 | 3219 | Silent Mutation | CCA,CCT | P,P 870 | NP_001122305.1 | |
NM_020899.3 | 3219 | Silent Mutation | CCA,CCT | P,P 870 | NP_065950.2 | |
XM_006721563.3 | 3219 | Silent Mutation | CCA,CCT | P,P 870 | XP_006721626.1 | |
XM_006721564.2 | 3219 | Silent Mutation | CCA,CCT | P,P 870 | XP_006721627.1 | |
XM_011523972.2 | 3219 | Silent Mutation | CCA,CCT | P,P 870 | XP_011522274.1 |