Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCGCCTCATCACTGGCATGGA[C/T]CGAGGCCTCATGGGCATGTGTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP10 - FK506 binding protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021939.3 | 634 | Silent Mutation | GAC,GAT | D,D 103 | NP_068758.3 | |
XM_011525099.2 | 634 | Silent Mutation | GAC,GAT | D,D 103 | XP_011523401.1 | |
XM_011525100.2 | 634 | Intron | XP_011523402.1 |
NT5C3B - 5'-nucleotidase, cytosolic IIIB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P3H4 - prolyl 3-hydroxylase family member 4 (non-enzymatic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |