Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGAGCTTGAAGCCTTGGCCCC[A/G]CCTTGGGCCTCCTCGCAGCCCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6883 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6883 - microRNA 6883 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PER1 - period circadian clock 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002616.2 | 3826 | Silent Mutation | GGC,GGT | G,G 1256 | NP_002607.2 | |
XM_005256689.1 | 3826 | Silent Mutation | GGC,GGT | G,G 1256 | XP_005256746.1 | |
XM_005256690.1 | 3826 | Silent Mutation | GGC,GGT | G,G 1196 | XP_005256747.1 |