Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCATTGTGGATTCTCCGAATG[A/G]CTTCAGAAAGAATCAAACTGATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPSAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPSAP1 - phosphoribosyl pyrophosphate synthetase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002766.2 | 1537 | Missense Mutation | GCC,GTC | A,V 364 | NP_002757.2 | |
XM_011525034.2 | 1537 | Missense Mutation | GCC,GTC | A,V 261 | XP_011523336.1 |
QRICH2 - glutamine rich 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |