Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTGAGCAGCAGCAGGCCACCCA[A/G]CACACCCACCAGCACCCACCAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607759 | ||||||||||||||||||||
Literature Links: |
ITGA2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGA2B - integrin subunit alpha 2b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000419.4 | 2943 | Silent Mutation | CTG,TTG | L,L 1005 | NP_000410.2 | |
XM_011524749.1 | 2943 | Silent Mutation | CTG,TTG | L,L 971 | XP_011523051.1 | |
XM_011524750.1 | 2943 | Intron | XP_011523052.1 |