Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGAGCTGTTTCACTGTGAGGGT[C/G]TTTATCCTTCATGCAGAGCTAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMIGD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMIGD1 - transmembrane and immunoglobulin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319942.1 | 862 | UTR 3 | NP_001306871.1 | |||
NM_206832.2 | 862 | Missense Mutation | CAC,GAC | H,D 255 | NP_996663.1 | |
XM_011524787.1 | 862 | Missense Mutation | CAC,GAC | H,D 255 | XP_011523089.1 |