Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGATATTGGTGCGGATGCAGC[A/G]GTTGTTGAGTGGCTGGACAGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA10 - carbonic anhydrase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082533.1 | 1869 | Missense Mutation | CGC,TGC | R,C 295 | NP_001076002.1 | |
NM_001082534.1 | 1869 | Missense Mutation | CGC,TGC | R,C 295 | NP_001076003.1 | |
NM_020178.4 | 1869 | Missense Mutation | CGC,TGC | R,C 295 | NP_064563.1 | |
XM_017024878.1 | 1869 | Missense Mutation | CGC,TGC | R,C 198 | XP_016880367.1 |