Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATTTCCAAGAGCCATCACTGTC[C/T]GTGAGACCTATCCAGGTATTGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108361 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASGR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASGR2 - asialoglycoprotein receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001181.4 | 941 | Silent Mutation | ACA,ACG | T,T 238 | NP_001172.1 | |
NM_001201352.1 | 941 | Silent Mutation | ACA,ACG | T,T 233 | NP_001188281.1 | |
NM_080912.3 | 941 | Silent Mutation | ACA,ACG | T,T 238 | NP_550434.1 | |
NM_080913.3 | 941 | Silent Mutation | ACA,ACG | T,T 214 | NP_550435.1 | |
NM_080914.2 | 941 | Silent Mutation | ACA,ACG | T,T 219 | NP_550436.1 | |
XM_005256648.2 | 941 | Silent Mutation | ACA,ACG | T,T 214 | XP_005256705.1 | |
XM_006721524.2 | 941 | Silent Mutation | ACA,ACG | T,T 175 | XP_006721587.2 | |
XM_006721526.1 | 941 | Silent Mutation | ACA,ACG | T,T 214 | XP_006721589.1 | |
XM_011523862.2 | 941 | Silent Mutation | ACA,ACG | T,T 337 | XP_011522164.2 | |
XM_011523863.2 | 941 | Silent Mutation | ACA,ACG | T,T 226 | XP_011522165.1 | |
XM_011523864.2 | 941 | Silent Mutation | ACA,ACG | T,T 245 | XP_011522166.1 | |
XM_011523865.1 | 941 | Silent Mutation | ACA,ACG | T,T 226 | XP_011522167.1 | |
XM_011523866.1 | 941 | Silent Mutation | ACA,ACG | T,T 226 | XP_011522168.1 | |
XM_017024651.1 | 941 | Silent Mutation | ACA,ACG | T,T 219 | XP_016880140.1 | |
XM_017024652.1 | 941 | Silent Mutation | ACA,ACG | T,T 238 | XP_016880141.1 | |
XM_017024653.1 | 941 | Silent Mutation | ACA,ACG | T,T 238 | XP_016880142.1 |