Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTCCCGGGGAGGCTCCCCTCTG[C/T]GCCTGTGGGCCTCGTCTCCTCGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600187 MIM: 601935 MIM: 615865 | ||||||||||||||||||||
Literature Links: |
EIF5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF5A - eukaryotic translation initiation factor 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPS2 - G protein pathway suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEURL4 - neuralized E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005408.1 | 4146 | Missense Mutation | CAC,CGC | H,R 1380 | NP_001005408.1 | |
NM_032442.2 | 4146 | Missense Mutation | CAC,CGC | H,R 1382 | NP_115818.2 |