Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCTGGGCCTACCCTTGCTCGGC[C/T]GCCAGAGCCGGTCCATCCCATGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609226 | ||||||||||||||||||||
Literature Links: |
MIR4525 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4525 - microRNA 4525 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB40B - RAB40B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006822.2 | 792 | Missense Mutation | CAG,CGG | Q,R 185 | NP_006813.1 | |
XM_005256334.4 | 792 | Missense Mutation | CAG,CGG | Q,R 125 | XP_005256391.1 | |
XM_005256335.3 | 792 | Intron | XP_005256392.1 | |||
XM_006722271.3 | 792 | Missense Mutation | CAG,CGG | Q,R 145 | XP_006722334.1 | |
XM_011523528.2 | 792 | Missense Mutation | CAG,CGG | Q,R 168 | XP_011521830.1 | |
XM_017024042.1 | 792 | Intron | XP_016879531.1 | |||
XM_017024043.1 | 792 | Missense Mutation | CAG,CGG | Q,R 125 | XP_016879532.1 |
WDR45B - WD repeat domain 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |