Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTGAGCAGCTGCCCCAGCCAG[A/G]CCCTGCAGCTGGTGAGTGTCAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138970 MIM: 607000 | ||||||||||||||||||||
Literature Links: |
CSF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSF3 - colony stimulating factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000759.3 | 406 | Missense Mutation | ACC,GCC | T,A 101 | NP_000750.1 | |
NM_001178147.1 | 406 | Intron | NP_001171618.1 | |||
NM_172219.2 | 406 | Missense Mutation | ACC,GCC | T,A 98 | NP_757373.1 | |
NM_172220.2 | 406 | Intron | NP_757374.2 |
MED24 - mediator complex subunit 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |