Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAACGTGCTCACCATGCTGGATG[A/G]GGCCTATATTCAGCCACAGCCTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609523 | ||||||||||||||||||||
Literature Links: |
ALDH3A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH3A2 - aldehyde dehydrogenase 3 family member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000382.2 | 410 | Missense Mutation | GAG,GGG | E,G 94 | NP_000373.1 | |
NM_001031806.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | NP_001026976.1 | |
XM_011523732.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_011522034.1 | |
XM_011523733.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_011522035.1 | |
XM_017024355.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_016879844.1 | |
XM_017024356.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_016879845.1 | |
XM_017024357.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_016879846.1 | |
XM_017024358.1 | 410 | Missense Mutation | GAG,GGG | E,G 94 | XP_016879847.1 |