Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGCAGGCGCGACAGTTTATAGG[C/T]GTCGTACTCCACTTGGTACTTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS7B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS7B - dehydrogenase/reductase 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM11 - transmembrane protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003876.2 | 861 | Missense Mutation | NP_003867.1 |