Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTCACCATGGTCTCAGCTCTG[C/T]GGGGAGCACCCCTGATCAGTGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612749 | ||||||||||||||||||||
Literature Links: |
SPACA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPACA3 - sperm acrosome associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317225.1 | 91 | Intron | NP_001304154.1 | |||
NM_001317226.1 | 91 | Missense Mutation | CGG,TGG | R,W 6 | NP_001304155.1 | |
NM_173847.4 | 91 | Missense Mutation | CGG,TGG | R,W 6 | NP_776246.1 |