Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAATATGTTTTATTTTTAATTAG[A/G]TCCAAACTAAATGACATGATAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGNBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGNBP2 - gametogenetin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024835.4 | 484 | Silent Mutation | GGA,GGG | G,G 143 | NP_079111.1 | |
XM_005257689.2 | 484 | Silent Mutation | GGA,GGG | G,G 143 | XP_005257746.1 | |
XM_011525265.2 | 484 | Silent Mutation | GGA,GGG | G,G 112 | XP_011523567.1 | |
XM_011525266.1 | 484 | Silent Mutation | GGA,GGG | G,G 143 | XP_011523568.1 | |
XM_017025107.1 | 484 | Silent Mutation | GGA,GGG | G,G 112 | XP_016880596.1 |