Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCCGCAGGGAGGGCAGAGGCTG[C/T]TGGCGGGTCAGGGAGGGCAGAGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186820 MIM: 602602 | ||||||||||||||||||||
Literature Links: |
CD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD7 - CD7 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006137.6 | 632 | Missense Mutation | ACA,GCA | T,A 175 | NP_006128.1 | |
XM_011523608.1 | 632 | Missense Mutation | ACA,GCA | T,A 75 | XP_011521910.1 | |
XM_017025316.1 | 632 | Missense Mutation | ACA,GCA | T,A 75 | XP_016880805.1 |
SECTM1 - secreted and transmembrane 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |