Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCCAGTGAGAAAATGGGATCCTG[C/G]CACCTGCCAAAAAGGGAAAGCGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602020 MIM: 607996 MIM: 602679 MIM: 606212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAFG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAFG - MAF bZIP transcription factor G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPB - neuropeptide B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCYT2 - phosphate cytidylyltransferase 2, ethanolamine | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT7 - sirtuin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016538.2 | 2662 | Missense Mutation | TGC,TGG | C,W 336 | NP_057622.1 | |
XM_006722284.2 | 2662 | Missense Mutation | TGC,TGG | C,W 254 | XP_006722347.1 | |
XM_011523580.1 | 2662 | Missense Mutation | TGC,TGG | C,W 301 | XP_011521882.1 | |
XM_011523581.2 | 2662 | Missense Mutation | TGC,TGG | C,W 265 | XP_011521883.1 |