Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTTTATGGTAAAACAGGCAGAG[A/C]ACATCTGCAGGCAAGCCACAATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604378 MIM: 614775 MIM: 601844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BECN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BECN1 - beclin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNTD1 - cyclin N-terminal domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173478.2 | 423 | Missense Mutation | AAC,CAC | N,H 90 | NP_775749.2 | |
XM_005257043.4 | 423 | Missense Mutation | AAC,CAC | N,H 84 | XP_005257100.1 | |
XM_005257044.4 | 423 | Missense Mutation | AAC,CAC | N,H 7 | XP_005257101.1 | |
XM_005257045.4 | 423 | Missense Mutation | AAC,CAC | N,H 7 | XP_005257102.1 | |
XM_006721690.2 | 423 | Missense Mutation | AAC,CAC | N,H 90 | XP_006721753.1 | |
XM_011524310.2 | 423 | Missense Mutation | AAC,CAC | N,H 7 | XP_011522612.1 | |
XM_011524311.2 | 423 | Missense Mutation | AAC,CAC | N,H 7 | XP_011522613.1 | |
XM_017024171.1 | 423 | Missense Mutation | AAC,CAC | N,H 7 | XP_016879660.1 |
COA3 - cytochrome c oxidase assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WNK4 - WNK lysine deficient protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |