Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCCCAGCCGCGCGCCCCGCGCC[C/T]CACGCCTGATGCTCTGTGCGCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGA3 - integrin subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002204.3 | 409 | Missense Mutation | CCA,TCA | P,S 12 | NP_002195.1 | |
XM_005257308.2 | 409 | Missense Mutation | CCA,TCA | P,S 12 | XP_005257365.1 |
LOC284080 - uncharacterized LOC284080 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |