Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGAGGCACCTGAAGAAGCGAGC[A/C]TGGTGTACGATGTTGTTTTCCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606735 | ||||||||||||||||||||
Literature Links: |
OSBPL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSBPL7 - oxysterol binding protein like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145798.2 | 2749 | Missense Mutation | CAG,CAT | Q,H 802 | NP_665741.1 | |
XM_017024122.1 | 2749 | Missense Mutation | CAG,CAT | Q,H 802 | XP_016879611.1 | |
XM_017024123.1 | 2749 | Missense Mutation | CAG,CAT | Q,H 802 | XP_016879612.1 | |
XM_017024124.1 | 2749 | Missense Mutation | CAG,CAT | Q,H 784 | XP_016879613.1 | |
XM_017024125.1 | 2749 | Missense Mutation | CAG,CAT | Q,H 364 | XP_016879614.1 |