Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGTCGCGCAGGAAGATCTGCCG[C/T]TTCTTGGCGATGGTGGTGGTGCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604330 | ||||||||||||||||||||
Literature Links: |
GRAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRAP - GRB2-related adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006613.3 | 658 | Silent Mutation | AAA,AAG | K,K 143 | NP_006604.1 | |
XM_017024029.1 | 658 | Silent Mutation | AAA,AAG | K,K 143 | XP_016879518.1 | |
XM_017024030.1 | 658 | Intron | XP_016879519.1 |
SLC5A10 - solute carrier family 5 member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |