Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCACCTCCCACTCGCTTCCCCA[C/T]CAGCCCAAAGAACTGGCTTGCCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607833 | ||||||||||||||||||||
Literature Links: |
FLJ45513 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLJ45513 - uncharacterized LOC729220 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAC4 - tachykinin 4 (hemokinin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077503.1 | 181 | Missense Mutation | ATG,GTG | M,V 61 | NP_001070971.1 | |
NM_001077504.1 | 181 | Missense Mutation | ATG,GTG | M,V 61 | NP_001070972.1 | |
NM_001077505.1 | 181 | Missense Mutation | ATG,GTG | M,V 61 | NP_001070973.1 | |
NM_001077506.1 | 181 | Missense Mutation | ATG,GTG | M,V 61 | NP_001070974.1 | |
NM_170685.2 | 181 | Missense Mutation | ATG,GTG | M,V 67 | NP_733786.2 |