Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTCCAGCCCAGGCACATGCCA[C/T]AAGCAACTCCAACAGCCAAGCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118955 MIM: 608625 MIM: 611753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLTC - clathrin heavy chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288653.1 | 156 | Intron | NP_001275582.1 | |||
NM_004859.3 | 156 | Intron | NP_004850.1 | |||
XM_005257012.3 | 156 | Intron | XP_005257069.1 |
PTRH2 - peptidyl-tRNA hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015509.2 | 156 | Missense Mutation | TAT,TGT | Y,C 26 | NP_001015509.1 | |
NM_016077.4 | 156 | Missense Mutation | TAT,TGT | Y,C 25 | NP_057161.1 | |
XM_011524887.1 | 156 | Missense Mutation | TAT,TGT | Y,C 25 | XP_011523189.1 |
VMP1 - vacuole membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |