Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAAGTAGGCTTTTATGTAAGTG[C/T]CATCAAGCTTCACTGCATTTGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601964 | ||||||||||||||||||||
Literature Links: |
DNAJC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC7 - DnaJ heat shock protein family (Hsp40) member C7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144766.2 | 1145 | Missense Mutation | GAC,GGC | D,G 269 | NP_001138238.1 | |
NM_003315.3 | 1145 | Missense Mutation | GAC,GGC | D,G 325 | NP_003306.3 | |
XM_011525167.2 | 1145 | Missense Mutation | GAC,GGC | D,G 269 | XP_011523469.1 | |
XM_011525168.2 | 1145 | Missense Mutation | GAC,GGC | D,G 269 | XP_011523470.1 | |
XM_011525169.2 | 1145 | Intron | XP_011523471.1 | |||
XM_017024994.1 | 1145 | Missense Mutation | GAC,GGC | D,G 269 | XP_016880483.1 |