Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTCTCTCCCTCCGCAGCTGGA[C/T]CTGTACGCGGGGGCTCTGTTTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM83G - family with sequence similarity 83 member G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039999.2 | 5025 | Intron | NP_001035088.2 | |||
XM_017024953.1 | 5025 | UTR 3 | XP_016880442.1 |
SLC5A10 - solute carrier family 5 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042450.2 | 5025 | Silent Mutation | GAC,GAT | D,D 153 | NP_001035915.1 | |
NM_001270648.1 | 5025 | Silent Mutation | GAC,GAT | D,D 153 | NP_001257577.1 | |
NM_001270649.1 | 5025 | Silent Mutation | GAC,GAT | D,D 153 | NP_001257578.1 | |
NM_001282417.1 | 5025 | Silent Mutation | GAC,GAT | D,D 97 | NP_001269346.1 | |
NM_152351.4 | 5025 | Silent Mutation | GAC,GAT | D,D 153 | NP_689564.3 | |
XM_011523652.1 | 5025 | Silent Mutation | GAC,GAT | D,D 195 | XP_011521954.1 | |
XM_011523653.2 | 5025 | Silent Mutation | GAC,GAT | D,D 195 | XP_011521955.1 | |
XM_017024191.1 | 5025 | Silent Mutation | GAC,GAT | D,D 195 | XP_016879680.1 |