Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGCTGCGGTAGGTTTGAATTT[T/C]ATTCTCCAGTCGGATCTTAATATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 148080 | ||||||||||||||||||||
Literature Links: |
KRT10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT10 - keratin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000421.3 | 1369 | Missense Mutation | NP_000412.3 | |||
XM_005257343.3 | 1369 | Nonsense Mutation | XP_005257400.1 |
TMEM99 - transmembrane protein 99 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195386.1 | 1369 | Intron | NP_001182315.1 | |||
NM_001195387.1 | 1369 | Intron | NP_001182316.1 | |||
NM_145274.3 | 1369 | Intron | NP_660317.2 | |||
XM_005257089.4 | 1369 | Intron | XP_005257146.1 | |||
XM_017024253.1 | 1369 | Intron | XP_016879742.1 |