Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGGACTTGACTTTCACTTGCAG[C/T]TTATCCACCAGATCCTGCAATCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYH3 - myosin, heavy chain 3, skeletal muscle, embryonic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002470.3 | 7068 | Silent Mutation | AAA,AAG | K,K 1871 | NP_002461.2 | |
XM_011523870.2 | 7068 | Silent Mutation | AAA,AAG | K,K 1871 | XP_011522172.1 | |
XM_011523871.2 | 7068 | Silent Mutation | AAA,AAG | K,K 1871 | XP_011522173.1 |
MYHAS - myosin heavy chain gene cluster antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |