Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCATGTCAGCCCCACTGGATGC[C/T]GTAAGTGAGGAGGAGGGAGTTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600735 MIM: 607526 MIM: 604631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFI35 - interferon induced protein 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005533.4 | 229 | Silent Mutation | GCC,GCT | A,A 7 | NP_005524.2 | |
XM_005257302.4 | 229 | Silent Mutation | GCC,GCT | A,A 7 | XP_005257359.1 | |
XM_017024584.1 | 229 | Intron | XP_016880073.1 | |||
XM_017024585.1 | 229 | Intron | XP_016880074.1 |
RPL27 - ribosomal protein L27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAT1 - vesicle amine transport 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |