Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAATTCTTCAACAAAAGAAGAGA[A/G]GGGAGAAGAAAAGCCTGACACCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606504 | ||||||||||||||||||||
Literature Links: |
CASC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASC3 - cancer susceptibility candidate 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007359.4 | 465 | Missense Mutation | AAG,AGG | K,R 131 | NP_031385.2 | |
XM_005257163.1 | 465 | Missense Mutation | AAG,AGG | K,R 131 | XP_005257220.1 |
MIR6866 - microRNA 6866 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |