Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGACAAGCTCACTGCTGAGAGT[C/T]GGTCTGGAGGACCCACTGGAGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610885 MIM: 614854 MIM: 611837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EME1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EME1 - essential meiotic structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC59 - leucine rich repeat containing 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018509.3 | 1146 | Missense Mutation | AAC,GAC | N,D 304 | NP_060979.2 |
MRPL27 - mitochondrial ribosomal protein L27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |