Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAAACTGAGCAAGTGGTTGATT[C/T]TTCAGATAGAGGTGAGTGGGTATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAT7 - lysine acetyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199155.1 | 545 | Missense Mutation | TCT,TTT | S,F 110 | NP_001186084.1 | |
NM_001199156.1 | 545 | Intron | NP_001186085.1 | |||
NM_001199157.1 | 545 | Missense Mutation | TCT,TTT | S,F 110 | NP_001186086.1 | |
NM_001199158.1 | 545 | Intron | NP_001186087.1 | |||
NM_007067.4 | 545 | Missense Mutation | TCT,TTT | S,F 110 | NP_008998.1 | |
XM_011524235.2 | 545 | Intron | XP_011522537.1 |