Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGAATAGGCTGTGTGCACCGCAT[A/G]CAGCACCCCCTTCTGGGCCATCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOS2 - nitric oxide synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000625.4 | 2976 | Missense Mutation | CAT,TAT | H,Y 1039 | NP_000616.3 | |
XM_011524859.2 | 2976 | Missense Mutation | CAT,TAT | H,Y 1039 | XP_011523161.1 | |
XM_011524860.2 | 2976 | Missense Mutation | CAT,TAT | H,Y 1038 | XP_011523162.1 | |
XM_011524861.1 | 2976 | Missense Mutation | CAT,TAT | H,Y 1015 | XP_011523163.1 | |
XM_011524862.1 | 2976 | Missense Mutation | CAT,TAT | H,Y 817 | XP_011523164.1 |