Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACGTCGAAAGCCTGATTCCTCA[C/T]CATCTGCATGAATCGGAGGCCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603704 | ||||||||||||||||||||
Literature Links: |
KRBA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRBA2 - KRAB-A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304947.1 | 2432 | Missense Mutation | ATG,GTG | M,V 312 | NP_001291876.1 | |
NM_213597.3 | 2432 | Missense Mutation | ATG,GTG | M,V 394 | NP_998762.1 | |
XM_005256443.2 | 2432 | Missense Mutation | ATG,GTG | M,V 360 | XP_005256500.1 | |
XM_011523643.1 | 2432 | Missense Mutation | ATG,GTG | M,V 360 | XP_011521945.1 | |
XM_011523644.1 | 2432 | Missense Mutation | ATG,GTG | M,V 360 | XP_011521946.1 | |
XM_017024163.1 | 2432 | Missense Mutation | ATG,GTG | M,V 360 | XP_016879652.1 | |
XM_017024164.1 | 2432 | Missense Mutation | ATG,GTG | M,V 357 | XP_016879653.1 | |
XM_017024165.1 | 2432 | Missense Mutation | ATG,GTG | M,V 312 | XP_016879654.1 |
LOC100128288 - uncharacterized LOC100128288 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL26 - ribosomal protein L26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |