Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGGGCTCTGGGGGCACTGTCTT[C/T]CGGCACTCACTCTGCAGCCGGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD15 - abhydrolase domain containing 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GIT1 - GIT ArfGAP 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085454.1 | 2365 | Silent Mutation | CGA,CGG | R,R 726 | NP_001078923.1 | |
NM_014030.3 | 2365 | Silent Mutation | CGA,CGG | R,R 717 | NP_054749.2 | |
XM_011524684.1 | 2365 | Silent Mutation | CGA,CGG | R,R 733 | XP_011522986.1 | |
XM_011524685.1 | 2365 | Silent Mutation | CGA,CGG | R,R 724 | XP_011522987.1 |
TP53I13 - tumor protein p53 inducible protein 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |